JWPH942F4082 J & W Pharmlab LLC

tert-butyl ((7S,8aR)-rel-2-benzyloctahydropyrrolo[1,2-a]pyrazin-7-yl)carbamate

  • MDL number MFCD32200742
  • Cas Number : 2216755-57-4
  • Molecular Weight : 331.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0886

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0886
Availability details for
Quantity
  • Availability
  • Add to Cart