JWPH942F4085 J & W Pharmlab LLC

(2R, 3S)-1-N-FMOC-3-Hydroxy-pyrrolidine-2-carboxylic acid

  • MDL number MFCD02682325
  • Cas Number : 1932810-08-6
  • Molecular Weight : 353.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0594

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0594
Availability details for
Quantity
  • Availability
  • Add to Cart