JWPH942F408B J & W Pharmlab LLC

8-Boc-hexahydro-2-oxa-5,8-diaza-cyclopenta[d]indene

  • MDL number MFCD17017576
  • Cas Number : 1251008-41-9
  • Molecular Weight : 268.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0600

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0600
Availability details for
Quantity
  • Availability
  • Add to Cart