JWPH942F4094 J & W Pharmlab LLC

1-Cbz-3-hydroxy-pyrrolidine-2-carboxylic acid

  • MDL number MFCD28398121
  • Cas Number : 57621-07-5
  • Molecular Weight : 265.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0614

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0614
Availability details for
Quantity
  • Availability
  • Add to Cart