JWPH942F409C J & W Pharmlab LLC

(3R,5S)-1-Benzyl-5-trifluoromethyl-pyrrolidine-3-carboxylic acid ethyl ester

  • MDL number MFCD29917143
  • Cas Number : 2381389-11-1
  • Molecular Weight : 301.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0642

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0642
Availability details for
Quantity
  • Availability
  • Add to Cart