JWPH942F40B3 J & W Pharmlab LLC

(1R,5S)-2-Aza-bicyclo[3.1.0]hexane-1-carboxylic acid methyl ester

  • MDL number MFCD19238040
  • Cas Number : 145631-97-6
  • Molecular Weight : 141.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0706

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0706
Availability details for
Quantity
  • Availability
  • Add to Cart