JWPH942F40D1 J & W Pharmlab LLC

Amino-(2-nitro-phenyl)-acetic acid ethyl ester

  • MDL number MFCD09997907
  • Cas Number : 791050-91-4
  • Molecular Weight : 224.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/78R0132

Safety Information

MSDS https://www.jwpharmlab.com/home/product/78R0132
Availability details for
Quantity
  • Availability
  • Add to Cart