JWPH942F40F9 J & W Pharmlab LLC

4-Amino-2-methyl-pyrrolidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD24467224
  • Cas Number : 1557656-80-0
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0955

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0955
Availability details for
Quantity
  • Availability
  • Add to Cart