JWPH942F410F J & W Pharmlab LLC

(3-Boc-amino-azetidin-1-yl)-furan-2-yl-acetic acid

  • MDL number MFCD04115176
  • Cas Number : 885276-12-0
  • Molecular Weight : 296.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/78R0038

Safety Information

MSDS https://www.jwpharmlab.com/home/product/78R0038
Availability details for
Quantity
  • Availability
  • Add to Cart