JWPH942F412E J & W Pharmlab LLC

4,4,8-Trimethyl-3,4-dihydro-1H-quinolin-2-one

  • MDL number MFCD12913838
  • Cas Number : 120689-98-7
  • Molecular Weight : 189.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/80R0210

Safety Information

MSDS https://www.jwpharmlab.com/home/product/80R0210
Availability details for
Quantity
  • Availability
  • Add to Cart