JWPH942F4161 J & W Pharmlab LLC

4-Chloro-6-methoxy-quinoline-3-carbonitrile

  • MDL number MFCD09261370
  • Cas Number : 13669-62-0
  • Molecular Weight : 218.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/80R0177

Safety Information

MSDS https://www.jwpharmlab.com/home/product/80R0177
Availability details for
Quantity
  • Availability
  • Add to Cart