JWPH942F4171 J & W Pharmlab LLC

2-(3-Amino-phenyl)-thiazole-4-carbaldehyde

  • MDL number MFCD06738373
  • Cas Number : 885278-99-9
  • Molecular Weight : 204.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/90R0055

Safety Information

MSDS https://www.jwpharmlab.com/home/product/90R0055
Availability details for
Quantity
  • Availability
  • Add to Cart