JWPH942F41BF J & W Pharmlab LLC

1,2,3,4-Tetrahydro-[1,8]naphthyridine-2-carboxylic acid

  • MDL number MFCD18257048
  • Cas Number : 1245913-51-2
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/80R0417

Safety Information

MSDS https://www.jwpharmlab.com/home/product/80R0417
Availability details for
Quantity
  • Availability
  • Add to Cart