JWPH942F41C3 J & W Pharmlab LLC

5-(4-Chloro-benzyl)-2H-tetrazole

  • MDL number MFCD00205200
  • Cas Number : 14064-61-0
  • Molecular Weight : 194.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/82R0006

Safety Information

MSDS https://www.jwpharmlab.com/home/product/82R0006
Availability details for
Quantity
  • Availability
  • Add to Cart