JWPH942F41F8 J & W Pharmlab LLC

3-Boc-2-(4-bromo-phenyl)-thiazolidine-4-carboxylic acid

  • MDL number MFCD04115281
  • Cas Number : 1008262-90-5
  • Molecular Weight : 388.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/90R0002

Safety Information

MSDS https://www.jwpharmlab.com/home/product/90R0002
Availability details for
Quantity
  • Availability
  • Add to Cart