JWPH942F420A J & W Pharmlab LLC

[2-(2-Methoxy-phenyl)-thiazol-4-yl]-methanol

  • MDL number MFCD06738391
  • Cas Number : 187793-07-3
  • Molecular Weight : 221.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/90R0195

Safety Information

MSDS https://www.jwpharmlab.com/home/product/90R0195
Availability details for
Quantity
  • Availability
  • Add to Cart