JWPH942F429E J & W Pharmlab LLC

cis-Diethyl pyrrolidine-2,5-dicarboxylate

  • MDL number MFCD01935388
  • Cas Number : 90514-00-4
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0970

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0970
Availability details for
Quantity
  • Availability
  • Add to Cart