JWPH942F429F J & W Pharmlab LLC

(2R,5R)-Diethyl pyrrolidine-2,5-dicarboxylate

  • MDL number MFCD28348118
  • Cas Number : 1808069-02-4
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0972

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0972
Availability details for
Quantity
  • Availability
  • Add to Cart