JWPH942F42C2 J & W Pharmlab LLC

6-Bromo-3-nitro-2-trifluoromethyl-pyridine

  • MDL number MFCD27923129
  • Cas Number : 1224321-68-9
  • Molecular Weight : 270.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/69R1432

Safety Information

MSDS https://www.jwpharmlab.com/home/product/69R1432
Availability details for
Quantity
  • Availability
  • Add to Cart