JWPH942F42CC J & W Pharmlab LLC

5-Bromo-3-nitro-2-trifluoromethyl-pyridine

  • MDL number MFCD18255263
  • Cas Number : 1211540-75-8
  • Molecular Weight : 270.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/69R1440

Safety Information

MSDS https://www.jwpharmlab.com/home/product/69R1440
Availability details for
Quantity
  • Availability
  • Add to Cart