JWPH942F42E8 J & W Pharmlab LLC

cis-2-Formyl-5-vinyl-pyrrolidine-1-carboxylic acid tert-butyl ester

  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0985

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0985
Availability details for
Quantity
  • Availability
  • Add to Cart