JWPH942F6280 J & W Pharmlab LLC

Decahydro-quinoxaline dihydrochloride

  • MDL number NA
  • Cas Number : 1082654-24-7
  • Molecular Weight : 213.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form Off White Solid
Shipping Temperature Room Temp
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/65R0199S

Safety Information

MSDS https://www.jwpharmlab.com/home/product/65R0199S
Availability details for
Quantity
  • Availability
  • Add to Cart