JWPH95E08DD0 J & W Pharmlab LLC

(2-Chloro-5-fluoro-pyridin-3-yl)-carbamic acid tert-butyl ester

  • MDL number MFCD22392295
  • Cas Number : 1394899-03-6
  • Molecular Weight : 246.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Room Temp
Physical Form Off White Solid
Shipping Temperature Room Temp
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/69R1472

Safety Information

MSDS https://www.jwpharmlab.com/home/product/69R1472
Availability details for
Quantity
  • Availability
  • Add to Cart