KEY001123381 Key Organics / BIONET

7-chloro-2,4-bis(trifluoromethyl)-1,8-naphthyridine

  • MDL number MFCD00140156
  • Cas Number : 106582-41-6
  • Molecular Weight : 300.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 92
Iupac Name 7-chloro-2,4-bis(trifluoromethyl)[1,8]naphthyridine
Inchi Code 1S/C10H3ClF6N2/c11-7-2-1-4-5(9(12,13)14)3-6(10(15,16)17)18-8(4)19-7/h1-3H
Inchi Key XLLWATLHAAVGDL-UHFFFAOYSA-N

Description

Melting Point 92
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/150

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/10D-005_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart