KEY001123536 Key Organics / BIONET

1-[(3,4-difluorophenyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid

  • MDL number MFCD01316311
  • Cas Number : 242797-29-1
  • Molecular Weight : 265.216

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214 - 216
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/317

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10E-320S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart