KEY001123583 Key Organics / BIONET

1-benzyl-2-oxo-1,2-dihydropyridine-3-carbohydrazide

  • MDL number MFCD00140469
  • Cas Number : 242472-15-7
  • Molecular Weight : 243.266

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110 - 114
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/374

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10E-393S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart