KEY001123615 Key Organics / BIONET

5-(4-aminophenyl)-5-methyl-2-phenyl-1,2,4-triazolidin-3-one

  • MDL number MFCD00140480
  • Cas Number : 245039-30-9
  • Molecular Weight : 268.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197 - 200
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/414

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10E-924_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart