KEY001123720 Key Organics / BIONET

[5-chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol

  • MDL number MFCD00141467
  • Cas Number : 282523-11-9
  • Molecular Weight : 214.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94 - 95 / 94-95@0.8mm Hg
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/531

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10F-341S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart