KEY001123731 Key Organics / BIONET

2-{[5-(1-benzyl-2-oxo-1,2-dihydropyridin-3-yl)-4-methyl-4H-1,2,4-triazol-3-yl]sulfanyl}propanoic acid

  • MDL number MFCD00141403
  • Cas Number : 282523-27-7
  • Molecular Weight : 370.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194 - 197
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/543

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10F-353S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart