KEY001123741 Key Organics / BIONET

5-(3-chlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD00243688
  • Cas Number : 282523-36-8
  • Molecular Weight : 304.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61 - 63
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/554

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10F-366S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart