KEY001123775 Key Organics / BIONET

1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperidine-4-carboxylic acid

  • MDL number MFCD00172245
  • Cas Number : 337919-65-0
  • Molecular Weight : 308.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135 - 138
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/594

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/10F-906_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart