KEY001123831 Key Organics / BIONET

3-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]sulfanyl}-1H-1,2,4-triazol-5-amine

  • MDL number MFCD00243559
  • Cas Number : 250714-16-0
  • Molecular Weight : 295.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 262 - 265
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/651

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10G-003_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart