KEY001123845 Key Organics / BIONET

2-(dimethylamino)-2-(1-methyl-1H-pyrrol-2-yl)acetonitrile

  • MDL number MFCD00231710
  • Cas Number : 117068-07-2
  • Molecular Weight : 163.224

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39 - 41 / 110@5mm Hg
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/665

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10G-025_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart