KEY001123870 Key Organics / BIONET

2-(4-chlorobenzenesulfonamido)-2-phenylacetic acid

  • MDL number MFCD00172800
  • Cas Number : 251096-79-4
  • Molecular Weight : 325.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175 - 177
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/695

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10G-322S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart