KEY001123876 Key Organics / BIONET

methyl 4-methyl-2-(4-methylbenzenesulfonamido)pentanoate

  • MDL number MFCD00664346
  • Cas Number : 51220-84-9
  • Molecular Weight : 299.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54 - 56.5
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/701

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10G-329S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart