KEY001123877 Key Organics / BIONET

3-{[(3,4-dichlorophenyl)methyl]sulfanyl}thiophene-2-carboxylic acid

  • MDL number MFCD00663856
  • Cas Number : 251096-84-1
  • Molecular Weight : 319.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189 - 190
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/702

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10G-330S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart