KEY001123919 Key Organics / BIONET

2-(4-fluorobenzenesulfonamido)-3-methylbutanoic acid

  • MDL number MFCD00664351
  • Cas Number : 250714-79-5
  • Molecular Weight : 275.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159 - 161
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/749

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10G-383S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart