KEY001123925 Key Organics / BIONET

3-methyl-2-[3-(trifluoromethyl)benzenesulfonamido]butanoic acid

  • MDL number MFCD00173010
  • Cas Number : 250714-85-3
  • Molecular Weight : 325.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127 - 129
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/761

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10G-397S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart