KEY001123932 Key Organics / BIONET

2-(4-fluorobenzenesulfonamido)-3-phenylpropanoic acid

  • MDL number MFCD00173011
  • Cas Number : 40279-96-7
  • Molecular Weight : 323.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108 - 110
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/783

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10G-421S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart