KEY001123933 Key Organics / BIONET

2-(4-bromobenzenesulfonamido)-3-phenylpropanoic acid

  • MDL number MFCD00173012
  • Cas Number : 37642-61-8
  • Molecular Weight : 384.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165 - 167
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/784

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10G-422S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart