KEY001123934 Key Organics / BIONET

2-(4-methoxybenzenesulfonamido)-3-phenylpropanoic acid

  • MDL number MFCD00173013
  • Cas Number : 40280-00-0
  • Molecular Weight : 335.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149 - 151
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/785

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10G-423S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart