KEY001123966 Key Organics / BIONET

2-(4-fluorobenzenesulfonamido)-4-methylpentanoic acid

  • MDL number MFCD00172812
  • Cas Number : 251097-25-3
  • Molecular Weight : 289.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124 - 126
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/826

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10G-480S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart