KEY001124000 Key Organics / BIONET

2-(4-bromobenzenesulfonamido)-4-methylpentanoic acid

  • MDL number MFCD00172816
  • Cas Number : 68305-78-2
  • Molecular Weight : 350.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128 - 130
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/863

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10G-521S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart