KEY001124023 Key Organics / BIONET

5-({[3-chloro-5-(trifluoromethyl)pyridin-2-yl]amino}methyl)-4,5-dihydro-1,2-oxazole-3-carbohydrazide

  • MDL number MFCD00202621
  • Cas Number : 251310-32-4
  • Molecular Weight : 337.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173 - 175
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/889

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10G-922_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart