KEY001124034 Key Organics / BIONET

1-(furan-2-carbonyl)-4-hydroxypyrrolidine-2-carboxylic acid

  • MDL number MFCD00202703
  • Cas Number : 251310-45-9
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224 - 227
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/903

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10G-942_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart