KEY001124062 Key Organics / BIONET

4-(4-fluorobenzoyl)-1-methyl-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD00172167
  • Cas Number : 128843-61-8
  • Molecular Weight : 231.226

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120 - 122
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/934

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10H-017_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart