KEY001124439 Key Organics / BIONET

2-(4-benzoylphenyl)-2-(4-fluorophenyl)acetonitrile

  • MDL number MFCD01316984
  • Cas Number : 446275-89-4
  • Molecular Weight : 315.347

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88 - 90
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1391

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10K-017_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart