KEY001124540 Key Organics / BIONET

2-[N-(4-chlorophenyl)benzenesulfonamido]acetic acid

  • MDL number MFCD00406209
  • Cas Number : 117309-41-8
  • Molecular Weight : 325.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161 - 162
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1498

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10K-376S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart