KEY001124568 Key Organics / BIONET

methyl 5-cyano-6-hydroxy-2-methylpyridine-3-carboxylate

  • MDL number MFCD00793171
  • Cas Number : 71408-02-1
  • Molecular Weight : 192.174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >250 DEC
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1531

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10K-419S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart