KEY001124620 Key Organics / BIONET

3-amino-4-(phenylsulfanyl)benzonitrile

  • MDL number MFCD00793304
  • Cas Number : 337923-85-0
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103 - 106
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1597

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10K-591S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart